Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIT1, Mouse, PE, Clone: SIT-01, Abnova™

Catalog No. 89111871 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-871 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-871 Supplier Abnova Corporation Supplier No. MAB4525
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SIT1.

Specifications

Antigen SIT1
Applications Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone SIT-01
Concentration 0.1 mg/mL
Conjugate PE
Description Mouse monoclonal antibody raised against partial recombinant SIT1.
Dilution Flow Cytometry (1:50) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene SIT1
Gene Alias MGC125908/MGC125909/MGC125910/RP11-331F9.5/SIT
Gene Symbols SIT1
Host Species Mouse
Immunogen Recombinant protein corresponding to partial human SIT1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27240
Test Specificity This antibody reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs. It weakly crossreacts with mouse SIT.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.