Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SIX1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105448 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-448 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-448 Supplier Abnova Corporation Supplier No. H00006495A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant SIX1.

The protein encoded by this gene is a homeobox protein that is similar to the Drosophila 'sine oculis' gene product. This gene is found in a cluster of related genes on chromosome 14 and is thought to be involved in limb development. Defects in this gene are a cause of autosomal dominant deafness type 23 (DFNA23) and branchiootic syndrome type 3 (BOS3). [provided by RefSeq]

Sequence: MSMLPSFGFTQEQVACVCEVLQQGGNLERLGRFLWSLPACDHLHKNESVLKAKAVVAFHRGNFRELYKILESHQFSPHNHPKLQQLWLKAHYVEAEKLCGRPLGAVGKYRVRRKFPLPRTIWDGEETSYCFKEKSRGVLREWYAHNPYPSPREKRELAEATGLTTTQVSNWFKNRRQRDRAAEAKERENTENNNSSSNKQNQLSPLEGGKPLMSSSEEEFSPPQSPDQNSVLLLQGNMGHARSSNYSLPGLTASQPSHGLQTHQHQLQDSLLGPLTSSLVDLGS

Specifications

Antigen SIX1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant SIX1.
Formulation 50% glycerol
Gene SIX1
Gene Accession No. BC008874
Gene Alias BOS3/DFNA23/TIP39
Gene Symbols SIX1
Host Species Mouse
Immunogen SIX1 (AAH08874, 1 a.a. ∽ 284 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6495
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.