Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-SKP1, Clone: 3F3.1,

Catalog No. MABS1229 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1229 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1229 Supplier MilliporeSigma™ Supplier No. MABS1229
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SKP1 clone: 3F3.1 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

S-phase kinase-associated protein 1 (UniProt P63208; also known as Cyclin-A/CDK2-associated protein p19, OCP-2, OCP-II, Organ of Corti protein 2, Organ of Corti protein II, p19A, p19skp1, RNA polymerase II elongation factor-like protein, SIII, Transcription elongation factor B polypeptide 1-like) is encoded by the SKP1 (also known as EMC19, OCP2, SKP1A, TCEB1L) gene (Gene ID 6500) in human. Polyubiquitination of proteins is mediated by ubiquitin-activating enzymes (E1s), ubiquitin-conjugating enzymes (E2s), and ubiquitin ligases (E3s). There exist RING finger-type, HECT-type, and U-box type of E3 ligases. Skp1 is one of the three core components of the SCF complex (Skp1, Cullin, F-box) and functions as an adapter that bridges the F-box protein and Cullin (CUL1). SCF complex depends on various F-box proteins for specific recognition and recruitment of substrates. SCF ligase plays an important role in regulating the transitions between G1/S and G2/M phases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SKP1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 3F3.1
Formulation Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P63208
Gene Symbols SKP1; EMC19; OCP2; SKP1A; TCEB1L
Host Species Mouse
Immunogen GST-tagged recombinant human SKP1 internal fragment.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_008861
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.