Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC12A6, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106188 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-188 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-188 Supplier Abnova Corporation Supplier No. H00009990A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SLC12A6.

This gene is a member of the K-Cl cotransporter (KCC) family. K-Cl cotransporters are integral membrane proteins that lower intracellular chloride concentrations below the electrochemical equilibrium potential. The proteins encoded by this gene are activated by cell swelling induced by hypotonic conditions. Alternate splicing results in multiple transcript variants encoding different isoforms. Mutations in this gene are associated with agenesis of the corpus callosum with peripheral neuropathy. [provided by RefSeq

Sequence: MPHFTVTKVEDPEEGAAASISQEPSLADIKARIQDSDEPDLSQNSITGEHSQLLDDGHKKARNAYLNNSNYEEGDEYFDKNLALFEEEMD

Specifications

Antigen SLC12A6
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC12A6.
Formulation 50% glycerol
Gene SLC12A6
Gene Accession No. NM_005135
Gene Alias ACCPN/DKFZp434D2135/KCC3/KCC3A/KCC3B
Gene Symbols SLC12A6
Host Species Mouse
Immunogen SLC12A6 (NP_005126, 1 a.a. ∽ 90 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9990
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.