Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc17a6, Mouse, PerCP, Clone: S29-29, Abnova™

Catalog No. 89109695 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-695 Supplier Abnova Corporation Supplier No. MAB11545
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant of Slc17a6.

Specifications

Antigen Slc17a6
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone S29-29
Conjugate PerCP
Description Mouse monoclonal antibody raised against recombinant of Slc17a6.
Dilution Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol)
Gene Slc17a6
Gene Alias Dnpi/Vglut2
Gene Symbols Slc17a6
Host Species Mouse
Immunogen Recombinant fusion protien corresponding to amino acids 501-582 at C-terminus of rat Slc17a6.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84487
Test Specificity Detects ∼60 kDa.
Target Species Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.