Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC1A3, Mouse, Clone: 3D1, Abnova™

Catalog No. 89105455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-455 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-455 Supplier Abnova Corporation Supplier No. H00006507M07A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC1A3.

Glutamate and aspartate are excitatory neurotransmitters that have been implicated in a number of pathologic states of the nervous system. Accumulation of extracellular excitatory amino acids can be cytotoxic and may also lower the seizure threshold in epilepsy. EAAT1 (SLC1A3) is a member of a family of high-affinity sodium-dependent transporter molecules that regulate neurotransmitter concentrations at the excitatory glutamatergic synapses of the mammalian central nervous system (Kirschner et al., 1994 [PubMed 8001975]).[supplied by OMIM

Sequence: VRVTAADAFLDLIRNMFPPNLVEACFKQFKTNYEKRSFKVPIQANETLVGAVINNVSEAMETLTRITEELVPVPGS

Specifications

Antigen SLC1A3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC1A3.
Formulation ascites with no preservative
Gene SLC1A3
Gene Accession No. NM_004172
Gene Alias EA6/EAAT1/FLJ25094/GLAST/GLAST1
Gene Symbols SLC1A3
Host Species Mouse
Immunogen SLC1A3 (NP_004163.2,162 a.a. ∽ 237 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6507
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.