Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC22A2, Mouse, Clone: 1E3, Abnova™

Catalog No. 89105463 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-463 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-463 Supplier Abnova Corporation Supplier No. H00006582M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SLC22A2.

Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. It is found primarily in the kidney, where it may mediate the first step in cation reabsorption. [provided by RefSeq

Sequence: HWRWLQFTVALPNFFFLLYYWCIPESPRWLISQNKNAEAMRIIKHIAKKNGKSLPASLQRLRLEEETGKKLNPSFLDLVRTPQIRKHTMILMYNWFTSSVLYQGLIKHMG

Specifications

Antigen SLC22A2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SLC22A2.
Formulation PBS with no preservative; pH 7.4
Gene SLC22A2
Gene Accession No. BC039899
Gene Alias MGC32628/OCT2
Gene Symbols SLC22A2
Host Species Mouse
Immunogen SLC22A2 (AAH39899,261 a.a. ∽ 370 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6582
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.