Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-SLC22A3, Clone: 4H6.1,

Catalog No. MABN359MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN359MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN359MI Supplier MilliporeSigma™ Supplier No. MABN359
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SLC22A3 clone: 4H6.1 in Human samples, and it is validated for Western Blotting

Solute carrier family 22 member 3 (UniProt O75751; also known as EMT, Extraneuronal monoamine transporterorganic cation transporter 3, Organic cation transporter 3) is encoded by the SLC22A3 (also known as EMTH, OCT3) gene (Gene ID 6581) in human. Solute carrier family 22 member 3 is a multi-transmembrane protein that mediates the outward transport and clearance of endogenous small organic cations as well as a wide array of drugs and environmental toxins. Known substrates for this transporter include histamine, serotonin, norepinephrine, dopamine and MPP+.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC22A3
Applications Western Blot
Classification Monoclonal
Clone 4H6.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O75751
Gene Symbols SLC22A3; EMTH; OCT3
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the C-terminal of human SLC22A3.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_068812
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.