Learn More
SLC26A3, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC26A3.
Supplier: Abnova Corporation H00001811A01
Description
The protein encoded by this gene is a transmembrane glycoprotein that transports chloride ions across the cell membrane in exchange for bicarbonate ions. It is localized to the mucosa of the lower intestinal tract, particularly to the apical membrane of columnar epithelium and some goblet cells. The protein is essential for intestinal chloride absorption, and mutations in this gene have been associated with congenital chloride diarrhea. [provided by RefSeq
Sequence: TQFPKCSTLANIGRTNIYKNKKDYYDMYEPEGVKIFRCPSPIYFANIGFFRRKLIDAVGFSPLRILRKRNKALRKIRKLQKQGLLQVTPKGFICTVDTSpecifications
| SLC26A3 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant SLC26A3. | |
| SLC26A3 | |
| CLD/DRA | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000111 | |
| SLC26A3 | |
| SLC26A3 (NP_000102, 503 a.a. ∽ 600 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 1811 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only