Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC2A10, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-383 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-383 Supplier Abnova Corporation Supplier No. H00081031A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SLC2A10.

SLC2A10 is a member of the facilitative glucose transporter family, which plays a significant role in maintaining glucose homeostasis.[supplied by OMIM

Sequence: SGPSCLAVPNATGQTGLPGDSGLLQDSSLPPIPRTNEDQREPILSTAKKTKPHPRSGD

Specifications

Antigen SLC2A10
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SLC2A10.
Formulation 50% glycerol
Gene SLC2A10
Gene Accession No. NM_030777
Gene Alias ATS/GLUT10/MGC126706
Gene Symbols SLC2A10
Host Species Mouse
Immunogen SLC2A10 (NP_110404, 325 a.a. ∽ 382 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81031
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.