Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC30A5, Mouse, Clone: 2E10, Abnova™

Catalog No. 89107300 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-300 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-300 Supplier Abnova Corporation Supplier No. H00064924M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SLC30A5.

Sequence: MEEKYGGDVLAGPGGGGGLGPVDVPSARLTKYIVLLCFTKFLKAVGLFESYDLLKAVHIVQFIFILKLGTAFFMVLFQKPFSSGKTITKHQIIGSLKIPGRKEFKDKKLNDPRKLVGN

Specifications

Antigen SLC30A5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SLC30A5.
Formulation ascites with no preservative
Gene SLC30A5
Gene Accession No. BC000808
Gene Alias FLJ12496/FLJ12756/MGC5499/ZNT5/ZNTL1/ZTL1/ZnT-5
Gene Symbols SLC30A5
Host Species Mouse
Immunogen SLC30A5 (AAH00808.1,1 a.a. ∽ 118 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64924
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.