Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC3A2, Mouse, Biotin, Clone: MEM-108, Abnova™

Catalog No. 89112070 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-070 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-070 Supplier Abnova Corporation Supplier No. MAB5040
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SLC3A2.

This gene is a member of the solute carrier family and encodes a cell surface, transmembrane protein with an alpha amylase domain. The protein exists as the heavy chain of a heterodimer, covalently bound through di-sulfide bonds to one of several possible light chains. It associates with integrins and mediates integrin-dependent signaling related to normal cell growth and tumorigenesis. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Specifications

Antigen SLC3A2
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-108
Conjugate Biotin
Description Mouse monoclonal antibody raised against native SLC3A2.
Dilution Flow Cytometry (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SLC3A2
Gene Alias 4F2/4F2HC/4T2HC/CD98/CD98HC/MDU1/NACAE
Gene Symbols SLC3A2
Host Species Mouse
Immunogen Native purified SLC3A2 from Raji Burkitt's lymphoma cell line.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6520
Test Specificity This antibody reacts with CD98, a 125 KDa disulfide-linked heterodimer (80 KDa glycosylated heavy chain + 45 KDa non-glykosylated light chain).
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.