Learn More
SLC4A4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC4A4.
Supplier: Abnova Corporation H00008671A01
Description
This gene encodes a sodium bicarbonate cotransporter (NBC) involved in the regulation of bicarbonate secretion and absorption and intracellular pH. Mutations in this gene are associated with proximal renal tubular acidosis. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MEKGSIMLDREASSLPQLVEMIVDHQIETGLLKPELKDKVTYTLLRKHRHQTKKSNLRSLADIGKTVSSASRMFTNPDNGSPAMTHRNLTSSSLNDISDKPEKDQLKNKSpecifications
SLC4A4 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SLC4A4. | |
SLC4A4 | |
DKFZp781H1314/HNBC1/KNBC/NBC1/NBC2/SLC4A5/hhNMC/pNBC | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003759 | |
SLC4A4 | |
SLC4A4 (NP_003750, 121 a.a. ∽ 229 a.a) partial recombinant protein with GST tag. | |
RUO | |
8671 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only