Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A5, Mouse, R-PE, Clone: FP5, Abnova™

Catalog No. 89109659 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-659 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-659 Supplier Abnova Corporation Supplier No. MAB11510
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant of SLC5A5.

This gene encodes a member of the sodium glucose cotransporter family. The encoded protein is responsible for the uptake of iodine in tissues such as the thyroid and lactating breast tissue. The iodine taken up by the thyroid is incorporated into the metabolic regulators triiodothyronine (T3) and tetraiodothyronine (T4). Mutations in this gene are associated with thyroid dyshormonogenesis 1

Specifications

Antigen SLC5A5
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone FP5
Conjugate R-PE
Description Mouse monoclonal antibody raised against recombinant of SLC5A5.
Dilution Immunofluorescence Immunohistochemistry Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol)
Gene SLC5A5
Gene Alias NIS
Gene Symbols SLC5A5
Host Species Mouse
Immunogen Recombinant mannose fusion protien corresponding to amino acids 468-643 of human SLC5A5.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6528
Test Specificity Mapped to amino acids 625-643 of hNIS. Apparent mol. wt of 97 kD, non-glycosylated version at 68 kD. Other minor bands associated with hNIS at 160 kDa, and degradation products at ∼30 kDa, and ∼15kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.