Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC5A7, Mouse, Clone: 62-2E8, Abnova™

Catalog No. 89110912 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-912 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-912 Supplier Abnova Corporation Supplier No. MAB2295
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SLC5A7.

Choline is a direct precursor of acetylcholine (ACh), a neurotransmitter of the central and peripheral nervous system that regulates a variety of autonomic, cognitive, and motor functions. SLC5A7 is a Na(+)- and Cl(-)- dependent high-affinity transporter that mediates the uptake of choline for acetylcholine synthesis in cholinergic neurons (Apparsundaram et al., 2000 [PubMed 11027560]).[supplied by OMIM

Specifications

Antigen SLC5A7
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 62-2E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SLC5A7.
Dilution Western Blot (1:500) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:500) Immunohistochemistry (Frozen sections) (1:500) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene SLC5A7
Gene Alias CHT/CHT1/MGC126299/MGC126300/hCHT
Gene Symbols SLC5A7
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of human SLC5A7.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60482
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.