Learn More
SLC6A20, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SLC6A20.
Supplier: Abnova Corporation H00054716A01
Description
Transport of small hydrophilic substances across cell membranes is mediated by substrate-specific transporter proteins which have been classified into several families of related genes. The protein encoded by this gene is a member of the subgroup of transporter with unidentified substrates within the Na+ and Cl- coupled transporter family. This gene is expressed in kidney, and its alternative splicing generates 2 transcript variants. [provided by RefSeq]
Sequence: KATFNYENCLKKVSLLLTNTFDLEDGFLTASNLEQVKGYLASAYPSKYSEMFPQIKNCSLESELDTAVQSpecifications
SLC6A20 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SLC6A20. | |
SLC6A20 | |
MGC161475/SIT1/XT3/Xtrp3 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_020208 | |
SLC6A20 | |
SLC6A20 (NP_064593, 301 a.a. ∽ 369 a.a) partial recombinant protein with GST tag. | |
RUO | |
54716 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only