Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc9a3, Mouse, Clone: 14D5, Abnova™

Catalog No. 89110911 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-911 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-911 Supplier Abnova Corporation Supplier No. MAB2294
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic phosphopeptide of Slc9a3.

Specifications

Antigen Slc9a3
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 14D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of Slc9a3.
Dilution Immunohistochemistry (1:50-1:1000) Immunofluorescence (1:50-1:1000) Western Blot (1-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene Slc9a3
Gene Alias Nhe3
Gene Symbols Slc9a3
Host Species Mouse
Immunogen Synthetic phosphopeptide corresponding to residues surrounding S552 of rat Slc9a3.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24784
Target Species Human, Mouse, Opossum, Rabbit, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.