Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC9A8, Mouse, Clone: 7A11, Abnova™

Catalog No. 89110909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-909 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-909 Supplier Abnova Corporation Supplier No. MAB2292
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SLC9A8.

Sodium-hydrogen exchangers (NHEs), such as SLC9A8, are integral transmembrane proteins that exchange extracellular Na+ for intracellular H+. NHEs have multiple functions, including intracellular pH homeostasis, cell volume regulation, and electroneutral NaCl absorption in epithelia (Xu et al., 2008 [PubMed 18209477]).[supplied by OMIM

Specifications

Antigen SLC9A8
Applications Western Blot
Classification Monoclonal
Clone 7A11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SLC9A8.
Dilution Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene SLC9A8
Gene Alias DKFZp686C03237/FLJ42500/KIAA0939/MGC138418/NHE8
Gene Symbols SLC9A8
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus of human SLC9A8.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23315
Test Specificity This antibody is specific to the 85 KDa mature glycosylated form of the protein. It does not recognize the 55 KDa immature form.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.