Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMAD4, Mouse, Clone: JM56, Abnova™

Catalog No. 89113719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-719 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-719 Supplier Abnova Corporation Supplier No. MAB9655

Mouse monoclonal antibody raised against full-length recombinant SMAD4.

Specifications

Antigen SMAD4
Applications Immunohistochemistry
Classification Monoclonal
Clone JM56
Conjugate Unconjugated
Description SMAD family member 4
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene SMAD4
Gene Alias DPC4, JIP, MADH4
Gene Symbols SMAD4
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human SMAD4.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4089
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.