Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMARCA2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105466 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-466 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-466 Supplier Abnova Corporation Supplier No. H00006595A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SMARCA2.

The protein encoded by this gene is a member of the SWI/SNF family of proteins and is highly similar to the brahma protein of Drosophila. Members of this family have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. Two transcript variants encoding different isoforms have been found for this gene, which contains a trinucleotide repeat (CAG) length polymorphism. [provided by RefSeq

Sequence: KILLDPNSEEVSEKDAKQIIETAKQDVDDEYSMQYSARGSQSYYTVAHAISERVEKQSALLINGTLKHYQLQGLEWM

Specifications

Antigen SMARCA2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SMARCA2.
Formulation 50% glycerol
Gene SMARCA2
Gene Accession No. NM_003070
Gene Alias BAF190/BRM/FLJ36757/MGC74511/SNF2/SNF2L2/SNF2LA/SWI2/Sth1p/hBRM/hSNF2a
Gene Symbols SMARCA2
Host Species Mouse
Immunogen SMARCA2 (NP_003061, 659 a.a. ∽ 735 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6595
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.