Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SMC1A, Mouse, Clone: C2M, Abnova™

Catalog No. 89110758 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-758 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-758 Supplier Abnova Corporation Supplier No. MAB2014
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SMC1A.

Proper cohesion of sister chromatids is a prerequisite for the correct segregation of chromosomes during cell division. The cohesin multiprotein complex is required for sister chromatid cohesion. This complex is composed partly of two structural maintenance of chromosomes (SMC) proteins, SMC3 and either SMC1L2 or the protein encoded by this gene. Most of the cohesin complexes dissociate from the chromosomes before mitosis, although those complexes at the kinetochore remain. Therefore, the encoded protein is thought to be an important part of functional kinetochores. In addition, this protein interacts with BRCA1 and is phosphorylated by ATM, indicating a potential role for this protein in DNA repair. This gene, which belongs to the SMC gene family, is located in an area of the X-chromosome that escapes X inactivation. [provided by RefSeq

Specifications

Antigen SMC1A
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone C2M
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SMC1A.
Dilution Western blot (1 to 5 ug/mL) Immunofluorescence (5 to 10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene SMC1A
Gene Alias CDLS2/DKFZp686L19178/DXS423E/KIAA0178/MGC138332/SB1.8/SMC1/SMC1L1/SMC1alpha/SMCB
Gene Symbols SMC1A
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 402-894 of human SMC1A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8243
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.