Learn More
SNFT, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SNFT.
Supplier: Abnova Corporation H00055509A01
Description
This gene encodes a member of the basic leucine zipper protein family. The encoded protein functions as a transcriptional repressor when heterodimerizing with JUN. The protein may play a role in repression of interleukin-2 and matrix metalloproteinase-1 transcription
Sequence: AAQRSRKKQTQKADKLHEEYESLEQENTMLRREIGKLTEELKHLTEALKEHEKMCPLLLCPMNFVPVPPRPDPVAGCLSpecifications
SNFT | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SNFT. | |
BATF3 | |
JDP1/JUNDM1/SNFT | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_018664 | |
BATF3 | |
SNFT (NP_061134, 48 a.a. to 125 a.a) partial recombinant protein with GST tag. | |
RUO | |
55509 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only