Learn More
SNX9, Mouse, Clone: 3C6, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SNX9.
Supplier: Abnova Corporation H00051429M03A
Description
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This protein does not contain a coiled coil region, like some family members, but does contain a SH3 domain near its N-terminus. This protein interacts with the cytoplasmic domains of the precursor but not the processed forms of a disintegrin and metalloprotease domain 9 and 15. This protein binds the beta-appendage domain of adaptor protein 2 and may function to assist adaptor protein 2 in its role at the plasma membrane. This protein interacts with activated Cdc42-associated kinase-2 to regulate the degradation of epidermal growth factor receptor protein. [provided by RefSeq
Sequence: MATKARVMYDFAAEPGNNELTVNEGEIITITNPDVGGGWLEGRNIKGERGLVPTDYVEILPSDGKDQFSCGNSVADQAFLDSLSASTAHASSSAASNNHQVGSGNDPWSAWSASKSGNWESSEGWGAQPEGAGAQRNTNTPNNWDTAFGHPQAYQGPATGDDDDWDEDWDGPKSSSYFKDSESADAGGAQRGNSRASSSSMKIPLNKFPGFAEPGTEQYLLAKQLAKPKEKIPIIVGDYGPMWVYPTSTFDCVVADPRKGSKMYGLKSYIEYQLTPTNTNRSVNHRYKHFDWLYERLLVKFGSAIPIPSLPDKQVTGRFEEEFIKMRMERLQAWMTRMCRHPVISESEVFQQFLNFRDEKEWKTGKRKAERDELAGVMIFSTMEPEAPDLDLVEIEQKCEAVGKFTKAMDDGVKELLTVGQEHWKRCTGPLPKEYQKIGKALQSLATVFSSSGYQGETDLNDAITEAGKTYEEIASLVAEQPKKDLHFLMECNHEYKGFLGCFPDVIGTHKGAIEKVKESDKLVATSKITLQDKQNMVKRVSIMSYALQAEMNHFHSNRIYDYNSVIRLYLEQQVQFYETIAEKLRQALSRFPVMSpecifications
SNX9 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC005022 | |
SNX9 | |
SNX9 (AAH05022,1 a.a. ∽ 595 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
51429 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3C6 | |
Mouse monoclonal antibody raised against a full-length recombinant SNX9. | |
SNX9 | |
MST155/MSTP155/SDP1/SH3PX1/SH3PXD3A/WISP | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.