Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1, Mouse, Clone: 18, Abnova™

Catalog No. 89113844 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-844 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-844 Supplier Abnova Corporation Supplier No. MAB9886
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant SOD1.

The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq

Specifications

Antigen SOD1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 18
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant SOD1.
Dilution Western blot (1:500) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene SOD1
Gene Accession No. NM_000454.4
Gene Alias ALS/ALS1/IPOA/SOD/homodimer
Gene Symbols SOD1
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human SOD1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6647
Test Specificity It can expression in HeLa whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.