Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD1, Mouse, Clone: 30F11, Abnova™

Catalog No. 89111448 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-448 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-448 Supplier Abnova Corporation Supplier No. MAB3499

Mouse monoclonal antibody raised against partial recombinant SOD1.

The protein encoded by this gene binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. The encoded isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene. [provided by RefSeq

Specifications

Antigen SOD1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 30F11
Conjugate Unconjugated
Description superoxide dismutase 1, soluble
Formulation Lyophilized from cell culture supernatant (0.09% sodium azide)
Gene SOD1
Gene Alias ALS, ALS1, IPOA, SOD, homodimer
Gene Symbols SOD1
Host Species Mouse
Immunogen Recombinant protein corresponding to regions I to V of human SOD1.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6647
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with distilled water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.