Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD2, Mouse, Clone: 23G5, Abnova™

Catalog No. 89108067 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-067 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-067 Supplier Abnova Corporation Supplier No. MAB0711

Mouse monoclonal antibody raised against partial recombinant SOD2.

This gene is a member of the iron/manganese superoxide dismutase family. It encodes a mitochondrial protein that forms a homotetramer and binds one manganese ion per subunit. This protein binds to the superoxide byproducts of oxidative phosphorylation and converts them to hydrogen peroxide and diatomic oxygen. Mutations in this gene have been associated with idiopathic cardiomyopathy (IDC), premature aging, sporadic motor neuron disease, and cancer. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. [provided by RefSeq

Specifications

Antigen SOD2
Applications Immunoprecipitation
Classification Monoclonal
Clone 23G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SOD2.
Dilution Immunoprecipitation (1-2 ul) The optimal working dilution should be determined by the end user.
Formulation In HEPES, 150mM NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide)
Gene SOD2
Gene Alias IPO-B/MNSOD/Mn-SOD
Gene Symbols SOD2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 25-222 of human SOD2.
Purification Method Ammonium sulfate precipitation
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6648
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.