Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOD3, Mouse, Clone: 4GG11G6, Abnova™

Catalog No. 89112971 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-971 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-971 Supplier Abnova Corporation Supplier No. MAB6644
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SOD3.

This gene encodes a member of the superoxide dismutase (SOD) protein family. SODs are antioxidant enzymes that catalyze the dismutation of two superoxide radicals into hydrogen peroxide and oxygen. The product of this gene is thought to protect the brain, lungs, and other tissues from oxidative stress. The protein is secreted into the extracellular space and forms a glycosylated homotetramer that is anchored to the extracellular matrix (ECM) and cell surfaces through an interaction with heparan sulfate proteoglycan and collagen. A fraction of the protein is cleaved near the C-terminus before secretion to generate circulating tetramers that do not interact with the ECM. [provided by RefSeq

Specifications

Antigen SOD3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4GG11G6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SOD3.
Dilution Western Blot (1:1000) Immunohistochemistry (1:100) Immunocytochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene SOD3
Gene Alias EC-SOD/MGC20077
Gene Symbols SOD3
Host Species Mouse
Immunogen Native purified SOD3 from human aortas.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6649
Test Specificity Detects EC SOD at ∼35KDa.
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.