Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-Somatotropin/GH1, Clone: 1H1.1,

Catalog No. MABC531MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC531MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC531MI Supplier MilliporeSigma™ Supplier No. MABC531
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Somatotropin/GH1 clone: 1H1.1 in Human samples, and it is validated for Western Blotting

Somatotropin (UniProt P01241; also known as GH, GH-N, Growth hormone 1, hGH-N, Pituitary growth hormone) is encoded by the GH1 (also known as GH, GHN, IGHD1B) gene (Gene ID 8856) in human. Somatotropin is a peptide hormone that plays an important role in growth control by inducing IGF-1 secretion in liver and other tissues. It stimulates myoblast differentiation and proliferation, as well as amino acids uptake and protein synthesis in muscle and other tissues. Acromegaly is a rare endocrine syndrome caused by a GH-secreting pituitary adenoma. Acromegaly may also be rarely associated with a hypothalamic or an ectopic GH-releasing hormone-producing tumor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Somatotropin/GH1
Applications Western Blot
Classification Monoclonal
Clone 1H1.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P01241
Gene Symbols GH1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human Somatotropin/GH1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_000506
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.