Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SORL1, Mouse, Clone: 7D7B11, Abnova™

Catalog No. 89108554 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-554 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-554 Supplier Abnova Corporation Supplier No. MAB10367
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SORL1.

This gene encodes a protein that belongs to the families of vacuolar protein sorting 10 (VPS10) domain-containing receptor proteins, of low density lipoprotein receptor (LDLR) proteins, and of fibronectin type III repeats proteins. In addition to VPS10, LDLR and fibronectin type 3 domains, this protein also includes an epidermal growth factor precursor-like module, a single transmembrane segment and a cytoplasmic tail with features similar to endocytosis- and sorting-competent receptors. Members of the VPS10 domain-containing receptor family are large with many exons but the CDS lengths are usually less than 3700 nt; this gene is an exception to the pattern with a CDS length greater than 6600 nt. Very large introns typically separate the exons encoding the VPS10 domain; the remaining exons are separated by much smaller-sized introns. The encoded protein is mainly intracellular and localizes in the paranuclear compartment. It is synthesized as a preproprotein, and when the propeptide is still attached, no binding occurs to the VPS10 domain. This gene is strongly expressed in the central nervous system. [provided by RefSeq

Specifications

Antigen SORL1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7D7B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SORL1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene SORL1
Gene Alias C11orf32/FLJ21930/FLJ39258/LR11/LRP9/SORLA/SorLA-1/gp250
Gene Symbols SORL1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 2159-2214 of human SORL1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6653
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.