Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOS1, Mouse, Clone: SOS-01, Abnova™

Catalog No. 89111515 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-515 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-515 Supplier Abnova Corporation Supplier No. MAB3649
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of SOS1.

This gene encodes a protein that is a guanine nucleotide exchange factor for RAS proteins, membrane proteins that bind guanine nucleotides and participate in signal transduction pathways. GTP binding activates and GTP hydrolysis inactivates RAS proteins. The product of this gene may regulate RAS proteins by facilitating the exchange of GTP for GDP. Mutations in this gene are associated with gingival fibromatosis 1 and Noonan syndrome type 4. [provided by RefSeq

Sequence: THPSMHRDGPPLLENAHSS

Specifications

Antigen SOS1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SOS-01
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of SOS1.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene SOS1
Gene Alias GF1/GGF1/GINGF/HGF/NS4
Gene Symbols SOS1
Host Species Mouse
Immunogen A synthetic peptide corresponding to human SOS1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6654
Test Specificity This antibody reacts with human SOS1, an ubiquitously expressed 150 KDa intracellular protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.