Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SOX18, Mouse, Clone: 2G12, Abnova™

Catalog No. 89106960 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-960 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-960 Supplier Abnova Corporation Supplier No. H00054345M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant SOX18.

This gene encodes a member of the SOX (SRY-related HMG-box) family of transcription factors involved in the regulation of embryonic development and in the determination of the cell fate. The encoded protein may act as a transcriptional regulator after forming a protein complex with other proteins. This protein plays a role in hair, blood vessel, and lymphatic vessel development. Mutations in this gene have been associated with recessive and dominant forms of hypotrichosis-lymphedema-telangiectasia. [provided by RefSeq

Sequence: EPGRYGLSPAGRGERQAADESRIRRPMNAFMVWAKDERKRLAQQNPDLHNAVLSKMLGKAWKELNAAEKRPFVEEAERLRVQHLRDHP

Specifications

Antigen SOX18
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2G12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant SOX18.
Formulation ascites with no preservative
Gene SOX18
Gene Accession No. NM_018419
Gene Alias HLTS
Gene Symbols SOX18
Host Species Mouse
Immunogen SOX18 (NP_060889,63 a.a. ∽ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54345
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.