Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-SOX9, Clone: 10E4.1,

Catalog No. MABC277 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC277 Supplier MilliporeSigma™ Supplier No. MABC277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SOX9 clone: 10E4.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation

Transcription factor SOX-9 (UniProt P48436; also known as Sex-determining region Y-box 9 protein, SRY-related HMG-box gene 9) is encoded by the SOX9 (also known as CMD1, CMPD1, SRA1) gene (Gene ID 6662) in human. Sox family of genes are related to the mammalian sex determining gene SRY. These genes contain sequences encoding for the HMG-box domains that mediate sequence-specific DNA binding. SOX genes encode transcriptional regulators involved in cell fate decision during development and the control of diverse developmental processes. The highly complex group of Sox genes cluster at at least 40 genomic loci and at least 30 Sox genes have been identified. SOX-9 is a key regulator of developmental processes, including male sex determination, chondrogenesis, neurogenesis, and neural crest development. SOX-9 up-regulation is reported in cancers of various origins, including breast, colon, prostate, and lung, where its expression correlates with malignancy and patient survival. Cellular SOX9 mRNA is shown to be targeted and negatively regulated by miR-32, and the cellular miR-32 level is reported to be significantly lower in non-small cell lung cancer (NSCLC) compared with noncancerous lung tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone 10E4.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P48436
Gene Symbols SOX9; CMD1; CMPD1; SRA1
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to a sequence from the N-terminal region of human SOX9.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_000337
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.