Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SP1, Mouse, Clone: 15H09, Abnova™

Catalog No. 89112942 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-942 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-942 Supplier Abnova Corporation Supplier No. MAB6565
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant SP1.

Specifications

Antigen SP1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 15H09
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant SP1.
Dilution Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1:50) Flow Cytometry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene SP1
Gene Symbols SP1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 150-200 of human SP1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6667
Test Specificity This affinity purified antibody recognizes ∼90 KDa of human SP1.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.