Learn More
SP110, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant SP110.
Supplier: Abnova Corporation H00003431A01
Description
The nuclear body is a multiprotein complex that may have a role in the regulation of gene transcription. This gene is a member of the SP100/SP140 family of nuclear body proteins and encodes a leukocyte-specific nuclear body component. The protein can function as an activator of gene transcription and may serve as a nuclear hormone receptor coactivator. In addition, it has been suggested that the protein may play a role in ribosome biogenesis and in the induction of myeloid cell differentiation. Alternative splicing has been observed for this gene and three transcript variants, encoding distinct isoforms, have been identified. [provided by RefSeq
Sequence: TPSDKKGKKRKRCIWSTPKRRHKKKSLPRGTASSRHGIQKKLKRVDQVPQKKDDSTCNSTVETRAQKARTECARKSRSEEIIDGTSEMNEGKRSQKTPSTPRRVTQGAASSpecifications
SP110 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant SP110. | |
SP110 | |
FLJ22835/IFI41/IFI75/IPR1/VODI | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_004510 | |
SP110 | |
SP110 (NP_004501, 271 a.a. ∽ 380 a.a) partial recombinant protein with GST tag. | |
RUO | |
3431 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only