Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPAG16, Mouse, Clone: 1D2, Abnova™

Catalog No. 89107325 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-325 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-325 Supplier Abnova Corporation Supplier No. H00079582M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SPAG16.

Cilia and flagella are comprised of a microtubular backbone, the axoneme, which is organized by the basal body and surrounded by plasma membrane. SPAG16 encodes 2 major proteins that associate with the axoneme of sperm tail and the nucleus of postmeiotic germ cells, respectively (Zhang et al., 2007 [PubMed 17699735]).[supplied by OMIM

Sequence: MAAQRGMPSSAVRVLEEALGMGLTAAGDARDTADAVAAEGAYYLEQVTITEASEDDYEYEEIPDDNFSIPEGEEDLAKAIQMAQEQATDTEILERKTVLPSKHAVPEVIEDFLCNFLIKMGMTRTLDCFQSEWYELIQKGVTELRTVGNVPDVYTQIMLLENENKNLKKDLKHYKQAAEYVIF

Specifications

Antigen SPAG16
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SPAG16.
Formulation PBS with no preservative; pH 7.4
Gene SPAG16
Gene Accession No. NM_001025436.1
Gene Alias DKFZp666P1710/FLJ22724/FLJ37717/MGC87036/PF20/WDR29
Gene Symbols SPAG16
Host Species Mouse
Immunogen SPAG16 (NP_001020607.1,1 a.a. ∽ 183 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79582
Target Species Human, Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.