Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARC, Mouse, Clone: ON1-1, Abnova™

Catalog No. 89113415 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-415 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-415 Supplier Abnova Corporation Supplier No. MAB8284
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against SPARC.

Specifications

Antigen SPARC
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone ON1-1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against SPARC.
Dilution ELISA (1-10 ug/mL) Western Blot (1-10 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS (1%BSA, 0.09% sodium azide)
Gene SPARC
Gene Alias MGC128090
Gene Symbols SPARC
Host Species Mouse
Immunogen Bovine SPARC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282077
Test Specificity This antibody reacts with bone-derived and platelet derived SPARC as well as with thrombin-stimulated platelets.
Target Species Bovine, Human, Pig, Rabbit
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized waterstore at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.