Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPARC, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89107904 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-904 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-904 Supplier Abnova Corporation Supplier No. MAB0278
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SPARC.

Secreted protein acidic and rich in cysteine/osteonectin/BM40, or SPARC, is a matrix-associated protein that elicits changes in cell shape, inhibits cell-cycle progression, and influences the synthesis of extracellular matrix (ECM) (Bradshaw et al., 2003 [PubMed 12721366]).[supplied by OMIM

Specifications

Antigen SPARC
Applications ELISA, Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SPARC.
Dilution Western Blot (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (50% glycerol)
Gene SPARC
Gene Alias ON
Gene Symbols SPARC
Host Species Mouse
Immunogen Native purified Human SPARC.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6678
Test Specificity This antibody recognizes human, rat and bovine bone osteonectin, human platelet osteonectin.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.