Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrin (broad spectrum), Mouse, Clone: RBC1/5B1, Abnova™

Catalog No. 89136670 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-670 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-670 Supplier Abnova Corporation Supplier No. MAB9509

Mouse monoclonal antibody raised against native spectrin (broad spectrum).

Specifications

Antigen Spectrin (broad spectrum)
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone RBC1/5B1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native spectrin (broad spectrum).
Dilution Immunohistochemistry (1:100) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Host Species Mouse
Immunogen Native spectrin (broad spectrum) from human red blood cell membrane.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Hamster, Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.