Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Spectrin, Clone: RBC2/3D5, Abnova™

Catalog No. 89136671 Shop All Abnova Corporation Products
Change view
Click to view available options
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
89-136-671 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-671 Supplier Abnova Corporation Supplier No. MAB9510

Specifications

Classification Monoclonal
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Primary or Secondary Primary
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.