Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPINK6, Mouse, Clone: 3C10, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant SPINK6.
Supplier: Abnova Corporation H00404203M04
Description
Sequence: MKLSGMFLLLSLALFCFLTGVFSQGGQVDCGEFQDTKVYCTRESNPHCGSDGQTYGNKCAFCKAIVKSGGKISLKHPGKCSpecifications
SPINK6 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC032003.1 | |
SPINK6 | |
SPINK6 (AAH32003.1,1 a.a. ∽ 80 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
3C10 | |
Mouse monoclonal antibody raised against a full-length recombinant SPINK6. | |
SPINK6 | |
BUSI2/MGC21394/UNQ844 | |
Mouse | |
Protein A Purification | |
RUO | |
404203 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction