Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPN, Mouse, Biotin, Clone: MEM-59, Abnova™

Catalog No. 89111889 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-889 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-889 Supplier Abnova Corporation Supplier No. MAB4548
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SPN.

Sialophorin (leukosialin) is a major sialoglycoprotein on the surface of human T lymphocytes, monocytes, granulocytes, and some B lymphocytes, which appears to be important for immune function and may be part of a physiologic ligand-receptor complex involved in T-cell activation.[supplied by OMIM

Specifications

Antigen SPN
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-59
Conjugate Biotin
Description Mouse monoclonal antibody raised against native SPN.
Dilution Flow Cytometry (1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.09% sodium azide)
Gene SPN
Gene Alias CD43/GPL115/LSN
Gene Symbols SPN
Host Species Mouse
Immunogen Native purified SPN from human T lymphocytes.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6693
Test Specificity This antibody recognizes neuraminidase-sensitive epitope on CD43 (Leukosialin), a 95-135 KDa type I transmembrane glycoprotein (mucin-type) which is involved in lymphocyte activation.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.