Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-SPOP, Clone: 9B7.1,

Catalog No. MABC565MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC565MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC565MI Supplier MilliporeSigma™ Supplier No. MABC565
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects SPOP clone: 9B7.1 in Human samples, and it is validated for Immunohistochemistry, Western Blotting

Speckle-type pox virus and zinc finger (POZ) protein, also known as HIB homolog 1 or Roadkill homolog 1, is encoded by the SPOP gene (also known as TEF2 or BTBD32) in human (Entrez Gene ID 8405). SPOP protein is a component of cullin-RING-based BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex and mediates recruitment of target proteins containing the SPOP-binding consensus (SBC) motif. The 42kDa SPOP protein comprises an N-terminal MATH domain, a bric-a-brac, tramtrack and broad complex (BTB)/POZ domain, a 3-box domain, and a C-terminal nuclear localization sequence. The MATH domain is primarily involved in substrate recognition/binding. Known substrates with SBC motifs include Macro H2A, Puc (Puckered), Daxx (Death domain–associated protein), and Gli (Glioma-associated oncogene). Prostate and endometrial cancers are frequently identified with SPOP mutations in the meprin and TRAF (Tumor necrosis factor receptor-associated factor) homology (MATH) domain, which can potentially lead to altered substrate binding activities. (PMID 25058385). The SPOP protein sequence is 100% conserved among human, murine, and rat (UniProt O43791, Q6ZWS8, and B2RYD9, respectively) species. Anti-SPOP, clone 9B7.1, is a purified mouse monoclonal antibody. A GST fusion protein containing the full-lenght human SPOP sequence was used as the immunogen. The binding epitope of clone 9B7.1 has not yet been mapped.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SPOP
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 9B7.1
Formulation Purified mouse monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. O43791
Gene Symbols SPOP
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human SPOP.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001007227
Target Species Human
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.