Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPP1, Mouse, Clone: 3G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant SPP1.
Supplier: Abnova Corporation H00006696M12A
Description
Sequence: SVVYGLRSKSKKFRRPDIQYPDATDEDITSHMESEELNGAYKAIPVAQDLNAPSDWDSRGKDSYETSQLDDQSAETHSHKQSRLYKRKANDESNEHSDVIDSQELSKVSRSpecifications
SPP1 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
NM_000582 | |
SPP1 | |
SPP1 (NP_000573,148 a.a. ∽ 257 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
6696 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
3G11 | |
Mouse monoclonal antibody raised against a partial recombinant SPP1. | |
SPP1 | |
BNSP/BSPI/ETA-1/MGC110940/OPN | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction