Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPPL2A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107471 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-471 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-471 Supplier Abnova Corporation Supplier No. H00084888A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SPPL2A.

This gene is a member of the signal peptide peptidase-like protease (SPPL) family and encodes an endosomal membrane protein with a protease associated (PA) domain. This protein plays a role in innate and adaptive immunity. A pseudogene of this gene also lies on chromosome 15. [provided by RefSeq

Sequence: HASGNGTTKDYCMLYNPYWTALPSTLENATSISLMNLTSTPLCNLSDIPPVGIKSKAVVVPWGSCHFLEKARIAQKGGAEAMLVVNNSVLFPPSGNRSE

Specifications

Antigen SPPL2A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SPPL2A.
Formulation 50% glycerol
Gene SPPL2A
Gene Accession No. NM_032802
Gene Alias IMP3/PSL2
Gene Symbols SPPL2A
Host Species Mouse
Immunogen SPPL2A (NP_116191, 31 a.a. ∽ 129 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84888
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.