Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRED2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107692 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-692 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-692 Supplier Abnova Corporation Supplier No. H00200734A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant SPRED2.

SPRED2 is a member of the Sprouty (see SPRY1; MIM 602465)/SPRED family of proteins that regulate growth factor-induced activation of the MAP kinase cascade (see MAPK1; MIM 176948) (Nonami et al., 2004 [PubMed 15465815]).[supplied by OMIM

Sequence: EDLIEGSTTSSSTIHNEAELGDDDVFTTATDSSSNSSQKREQPTRTISSPTSCEHRRIYTLGHLHDSYPTDHYHLDQPMPRPCRQVSFPDDDEEIVRINP

Specifications

Antigen SPRED2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant SPRED2.
Formulation 50% glycerol
Gene SPRED2
Gene Accession No. NM_181784
Gene Alias FLJ21897/FLJ31917/MGC163164/Spred-2
Gene Symbols SPRED2
Host Species Mouse
Immunogen SPRED2 (NP_861449, 120 a.a. ∽ 219 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 200734
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.