Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTAN1, Mouse, Clone: AA6, Abnova™

Catalog No. 89111043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-043 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-043 Supplier Abnova Corporation Supplier No. MAB2508
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SPTAN1.

Specifications

Antigen SPTAN1
Applications Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone AA6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SPTAN1.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene SPTAN1
Gene Alias SPECA
Gene Symbols SPTAN1
Host Species Mouse
Immunogen Native purified chicken SPTAN1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374234
Test Specificity This antibody is specific to the 240 kD alpha-fodrin molecule of all mammalian nonerythroid cells and chicken alpha-spectrin.
Target Species Chicken
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.