Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTB, Mouse, Clone: DB2, Abnova™

Catalog No. 89111049 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-049 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-049 Supplier Abnova Corporation Supplier No. MAB2516
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native SPTB.

Specifications

Antigen SPTB
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone DB2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native SPTB.
Dilution Western Blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene SPTB
Gene Alias HSpTB1
Gene Symbols SPTB
Host Species Mouse
Immunogen Native purified human SPTB.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6710
Test Specificity This antibody is specific to the 220kD human erythroid beta-spectrin. The protein is also present in the membranes of human skeletal muscle cells. Erythroid spectrins, some other proteins of erythroid cytoskeleton, and the transmembrane protein band 3 are highly specific to erythrocytes and their progenitors. They are more reliable markers for erythroid differentiation than Glycophorin A, the commonly used marker for erythroid differentiation, because Glycophorin A is expressed also in many cell lines otherwise exhibiting mainly megakaryotic charasteristics. Monoclonal antibody to erythroid beta-spectrin is derived from the hybridoma produced by fusion between myeloma cells and Balb/c spleen cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.