Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRC, Mouse, Clone: 28, Abnova™

Catalog No. 89110764 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-764 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-764 Supplier Abnova Corporation Supplier No. MAB2021

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against synthetic peptide of SRC.

This gene is highly similar to the v-src gene of Rous sarcoma virus. This proto-oncogene may play a role in the regulation of embryonic development and cell growth. The protein encoded by this gene is a tyrosine-protein kinase whose activity can be inhibited by phosphorylation by c-SRC kinase. Mutations in this gene could be involved in the malignant progression of colon cancer. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen SRC
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 28
Conjugate Unconjugated
Description v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian)
Formulation In PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Gene SRC
Gene Alias ASV, SRC1, c-SRC, p60-Src
Gene Symbols SRC
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 519-536 of human SRC.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6714
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.