Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SRCRB4D, Mouse, Clone: 3E9, Abnova™

Catalog No. 89107622 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-622 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-622 Supplier Abnova Corporation Supplier No. H00136853M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant SRCRB4D.

The scavenger receptor cysteine-rich (SRCR) superfamily is an ancient and highly conserved group of cell surface and/or secreted proteins, some of which are involved in the development of the immune system and the regulation of both innate and adaptive immune responses. Group B SRCR domains usually contain 8 regularly spaced cysteines that give rise to a well-defined intradomain disulfide-bond pattern.[supplied by OMIM

Sequence: MTGTLRTRAWPAAKRAAGLRWALRDEARLSDCFHLGWGQHNCGHHEDAGALCAGEADSEGPEELGLQVQQDGSETTRVPTPRPRDGHLRLVNGAHRCEGRVELYLGQRWGTVCDDAWDLRAAGVLCRQLGCGQALAAPGEAHFGPGRGPILLDNVKCRGEESALLLCSHIRWDAHNCDHSEDASVLCQPS

Specifications

Antigen SRCRB4D
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant SRCRB4D.
Formulation ascites with no preservative
Gene SRCRB4D
Gene Accession No. BC015651
Gene Alias S4D-SRCRB/SRCRB-S4D
Gene Symbols SRCRB4D
Host Species Mouse
Immunogen SRCRB4D (AAH15651.1,1 a.a. ∽ 190 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 136853
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.