Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SSSCA1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106345 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-345 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-345 Supplier Abnova Corporation Supplier No. H00010534A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant SSSCA1.

This antigen is recognized by a subset of anti-centromere antibodies from patients with scleroderma and/or Sjogren's syndrome. Subcellular localization has not yet been established. [provided by RefSeq]

Sequence: MALNGAEVDDFSWEPPTEAETKVLQARRERQDRISRLMGDYLLRGYRVLGETCADCGTILLQDKQRKIYCVACQELDSDVDKDNPALNAQAALSQAREHQLASASELPLGSRPAPQPPVPRPEHCEGAAAGLKAAQGPPAPAVPPNTDVMACTQTALLQKLTWASAELGSSTSLETSIQLCGLIRACAEALRSLQQLQH

Specifications

Antigen SSSCA1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant SSSCA1.
Formulation 50% glycerol
Gene SSSCA1
Gene Accession No. BC014791
Gene Alias p27
Gene Symbols SSSCA1
Host Species Mouse
Immunogen SSSCA1 (AAH14791.1, 1 a.a. ∽ 199 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10534
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.